Pyrogallol, Crystal, Reagent, ACS, Spectrum™ Chemical
P1460, 87-66-1, C6H3(OH)3
Supplier:  Spectrum Chemical Mfg Cor P1460125GMCS6
 View more versions of this product
Catalog No. 18606012
Explore available promotions
Description
Description
Spectrum™ Chemical Pyrogallol, Crystal, Reagent, ACS , also known as Pyrogallic acid, is one of three isomeric benzenetriols that is used for oxygen absorption in gas analysis as well as in hair dying. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Specifications
Specifications
87-66-1
131°C to 135°C
Amber Glass Bottle
MFCD00002192
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
1
5e-005
C6H6O3
6 x 125 g
OC1=CC=CC(O)=C1O
126.11
Chemical Identifiers
87-66-1
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
OC1=CC=CC(O)=C1O
C6H6O3
MFCD00002192
benzene-1,2,3-triol
Product Suggestions
Product Suggestions