missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrogallol (Crystalline/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLA263500

 View more versions of this product

Catalog No. A263-500



Explore available promotions

Description

Description

Specifications

Specifications

Pyrogallol
131°C
1.453g/cm³
309°C
131°C to 135°C
C6H6O3
MFCD00002192
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
OC1=CC=CC(O)=C1O
126.11
CHEBI:16164
Certified ACS
87-66-1
White
5.8
0.005% max.
Poly Bottle
C6H3-1, 2, 3-(OH)3
500 g
WQGWDDDVZFFDIG-UHFFFAOYSA-N
benzene-1,2,3-triol
1057
126.11
Powder or Solid

Chemical Identifiers

87-66-1
126.11
WQGWDDDVZFFDIG-UHFFFAOYSA-N
1057
benzene-1,2,3-triol
C6H6O3
MFCD00002192
pyrogallol, 1,2,3-trihydroxybenzene, pyrogallic acid, 1,2,3-benzenetriol, fouramine brown ap, fourrine pg, fourrine 85, pyro, c.i. oxidation base 32, piral
CHEBI:16164
OC1=CC=CC(O)=C1O
Videos
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 6.1, : Poison

SDS Documents

Documents

Promotions

Promotions