Learn More
Pyromellitic dianhydride, 98%
CAS: 89-32-7 | C10H2O6 | 218.12 g/mol
$136.45 - $1809.44
Chemical Identifiers
| CAS | 89-32-7 |
|---|---|
| Molecular Formula | C10H2O6 |
| Molecular Weight (g/mol) | 218.12 |
| MDL Number | MFCD00005005 |
| InChI Key | ANSXAPJVJOKRDJ-UHFFFAOYSA-N |
| Synonym | pyromellitic dianhydride, pyromellitic anhydride, pyromellitic acid anhydride, 1,2,4,5-benzenetetracarboxylic anhydride, pyromellitic acid dianhydride, 1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, pmda, benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetraone, unii-2z22t30qzq, 1,2,4,5-benzenetetracarboxylic dianhydride |
| PubChem CID | 6966 |
| SMILES | O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4578822
|
Thermo Scientific Chemicals
04578822 |
100 g |
Each for $136.45
|
|
|||||
|
AA4578836
|
Thermo Scientific Chemicals
04578836 |
500 g |
Each for $515.47
|
|
|||||
|
AA45788A3
|
Thermo Scientific Chemicals
045788A3 |
2 kg |
Each for $1,809.44
|
|
|||||
Description
Pyromellitic dianhydride is used as a monomer in the preparation of polyimide polymers such as kapton; thermoplastics such as polyesters, polyethers, plasticizers, and expoxy resins. It is involved in the preparation of high performance coatings due to its mechanical, thermal and oxidative stability properties. It acts as a curing agent for epoxy resins which find application in adhesives, coatings and molding powders. As an intermediate, it plays an important role in polyimide based materials like printed circuit boards, tape automated bonding and magnetic wire insulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 89-32-7 | |
| 218.12 | |
| ANSXAPJVJOKRDJ-UHFFFAOYSA-N | |
| 6966 |
| C10H2O6 | |
| MFCD00005005 | |
| pyromellitic dianhydride, pyromellitic anhydride, pyromellitic acid anhydride, 1,2,4,5-benzenetetracarboxylic anhydride, pyromellitic acid dianhydride, 1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, pmda, benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetraone, unii-2z22t30qzq, 1,2,4,5-benzenetetracarboxylic dianhydride | |
| O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O |
Specifications
| 89-32-7 | |
| White to Yellow | |
| 305°C to 310°C (30 mmHg) | |
| C10H2O6 | |
| 100 g | |
| Moisture sensitive | |
| Soluble in dimethyl sulfoxide,acetone,chloroform,ethyl ether,n-hexane and benzene. | |
| O=C1OC(=O)C2=CC3=C(C=C12)C(=O)OC3=O | |
| 218.12 | |
| 218.12 | |
| Crystalline Powder |
| 284°C to 288°C | |
| 1.68 g/mL | |
| 380°C (716°F) | |
| MFCD00005005 | |
| 213583 | |
| pyromellitic dianhydride, pyromellitic anhydride, pyromellitic acid anhydride, 1,2,4,5-benzenetetracarboxylic anhydride, pyromellitic acid dianhydride, 1h,3h-benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetrone, pmda, benzo 1,2-c:4,5-c' difuran-1,3,5,7-tetraone, unii-2z22t30qzq, 1,2,4,5-benzenetetracarboxylic dianhydride | |
| ANSXAPJVJOKRDJ-UHFFFAOYSA-N | |
| furo[3,4-f][2]benzofuran-1,3,5,7-tetrone | |
| 6966 | |
| 98% | |
| Pyromellitic dianhydride |
Safety and Handling
GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P272-P280-P285-P302+P352-P304+P341-P305+P351+P338-P310-P333+P313-P342+P311-P363-P501c
H317-H318-H334
EINECSNumber : 201-898-9
RTECSNumber : DB9300000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only