missing translation for 'onlineSavingsMsg'
Learn More

Pyronin Y

Marker dye for RNA useful in acid buffer systems | CAS: 92-32-0 | C17H19ClN2O | 302.802 g/mol

Supplier:  Thermo Scientific Chemicals J6106803

 View more versions of this product

Catalog No. AAJ6106803

Only null left
Explore available promotions

Description

Description

Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyronin Y is a cationic dye that intercalates RNA and is visible as a red band during electrophoresis. It has also been shown to accumulate in mitochondria of viable cells. In conjuction with Methyl Green or Nancy-520, Pyronin Y can be used as a fluorescent differential stain of nucleic acids in paraffin tissue sections.

Solubility
Soluble in water. Also soluble in methyl cellosolve (9 mg/mL) and ethanol (3 mg/mL).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

Pyronin Y
92-32-0
MFCD00011725
14,8007
Soluble in water. Also soluble in methyl cellosolve (9mg/mL) and ethanol (3mg/mL).
CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]
302.802
CHEBI:87347
Green
Crystalline Powder
250°C to 260°C
C17H19ClN2O
3795789
C.I. 45005; Pyronin G
INCIMLINXXICKS-UHFFFAOYSA-M
[6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
7085
302.8
1 g

Chemical Identifiers

92-32-0
302.802
INCIMLINXXICKS-UHFFFAOYSA-M
7085
[6-(dimethylamino)xanthen-3-ylidene]-dimethylazanium;chloride
C17H19ClN2O
MFCD00011725
C.I. 45005; Pyronin G
CHEBI:87347
CN(C)C1=CC2=C(C=C1)C=C3C=CC(=[N+](C)C)C=C3O2.[Cl-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P281-P308+P313-P501c

H341

EINECSNumber : 202-147-8

RTECSNumber : BQ1450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only