FUJIFILM Wako PYROSTAR™ ES-F 200 Test Kit
Endotoxin-specific and specially formulated to be unreactive to (1,3)- ß-D-glucan, Multi-test configuration designed for use with water bath, heat block or Toxinometer™
Supplier:  FUJIFILM Wako WPEK450006
 View more versions of this product
Catalog No. 12-333-007
Explore available promotions
Description
Description
  • Endotoxin-specific reagent avoids false positive results from glucans
  • Dual-purpose reagent can perform either gel clot or kinetic turbidimetric assay (KTA)
  • Gel clot sensitivities range from 0.015 to 0.25 EU/mL
  • Kinetic turbidimetric sensitivities range from 0.001 to 10 EU/mL
  • Chances of interference are reduced due to high sensitivity
  • Facilitates easier transition from gel clot to KTA
Specifications
Specifications
Lyophilized
4 x 5.2 mL vials of lysate and 1 vial of CSE (500 ng/vial)
0.06 EU/mL
5.2 mL
200 Tests