missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrole-2-carboxaldehyde, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 132061000

 View more versions of this product

Catalog No. AC132061000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Pyrrole-2-carboxaldehyde
1003-29-8
Authentic
Glass bottle
MFCD00005217
21, 270
ZSKGQVFRTSEPJT-UHFFFAOYSA-N
1H-pyrrole-2-carbaldehyde
13854
95.1
99%
106°C
98.5% min. (GC)
C5H5NO
100g
pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde
C1=CNC(=C1)C=O
95.1
CHEBI:59978
99%

Chemical Identifiers

1003-29-8
95.1
ZSKGQVFRTSEPJT-UHFFFAOYSA-N
13854
1H-pyrrole-2-carbaldehyde
C5H5NO
MFCD00005217
pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde
CHEBI:59978
C1=CNC(=C1)C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-705-5

TSCA : TSCA

Recommended Storage : product takes up color on ageing

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only