missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole, 99%, extra pure

CAS: 109-97-7 | C4H5N | 67.09 g/mol

$76.90 - $566.85

Chemical Identifiers

CAS 109-97-7
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00005216
InChI Key KAESVJOAVNADME-UHFFFAOYSA-N
Synonym pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
PubChem CID 8027
ChEBI CHEBI:19203
IUPAC Name 1H-pyrrole
SMILES N1C=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC157710250
SDS View Documents
Thermo Scientific Chemicals
157710250
25 g Glass bottle
Each for $76.90
Only null left
 
AC157711000
SDS View Documents
Thermo Scientific Chemicals
157711000
100 g Glass bottle
Each for $171.10
Case of 6 Each for $1,025.88
Only null left
 
AC157715000
SDS View Documents
Thermo Scientific Chemicals
157715000
500 g Glass bottle
Each for $566.85
Case of 6 Each for $0.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-97-7
67.09
KAESVJOAVNADME-UHFFFAOYSA-N
8027
1H-pyrrole
C4H5N
MFCD00005216
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
CHEBI:19203
N1C=CC=C1

Specifications

109-97-7
Brown to Colorless
129.0°C to 131.0°C
Authentic
Glass bottle
1.5080 to 1.5100
25 g
0.96
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
KAESVJOAVNADME-UHFFFAOYSA-N
1H-pyrrole
8027
CHEBI:19203
99%
Liquid
-23.0°C
0.9600g/mL
36°C
98.5% min. (GC)
C4H5N
MFCD00005216
20,159
15,8128
Solubility in water: 60g/L (20°C). Other solubilities: insoluble in dilute alkalies, soluble in alcohol, ether and dilute acids, soluble in most organic solvents
N1C=CC=C1
67.09
1.317 mPa.s (20°C)
67.09
Extra Pure
Pyrrole
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Causes serious eye damage.
Toxic if swallowed.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 203-724-7

RTECSNumber : UX9275000

TSCA : TSCA

Recommended Storage : Darkens on storage

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only