missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrolidine-3-carboxylic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 459110050

 View more versions of this product

Catalog No. AC459110050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Pyrrolidine-3-carboxylic acid
50.86 to max. 53.47% min. Carbon 11.86 to max. 12.47% min. Nitrogen
C5H9NO2
3-pyrrolidinecarboxylic acid, 3-pyrrolidine carboxylic acid, beta-proline, 3-pyrrolidinecarboxylicacid, pyrrolidine-3-carboxylicacid, dl-pyrrolidine-3-carboxylic acid, beta-dl-proline, pubchem10439, acmc-209mcv, acmc-20a9vh
C1CNCC1C(=O)O
115.13
115.13
59378-87-9
Authentic
5g
JAEIBKXSIXOLOL-UHFFFAOYSA-N
pyrrolidine-3-carboxylic acid
3034645
98%

Chemical Identifiers

59378-87-9
115.13
3-pyrrolidinecarboxylic acid, 3-pyrrolidine carboxylic acid, beta-proline, 3-pyrrolidinecarboxylicacid, pyrrolidine-3-carboxylicacid, dl-pyrrolidine-3-carboxylic acid, beta-dl-proline, pubchem10439, acmc-209mcv, acmc-20a9vh
pyrrolidine-3-carboxylic acid
C5H9NO2
JAEIBKXSIXOLOL-UHFFFAOYSA-N
3034645
C1CNCC1C(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye damage.

GHS P Statement Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only