missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyrrolidine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A14852AE

 View more versions of this product

Catalog No. AAA14852AE


Explore available promotions

Description

Description

Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.

Solubility
Miscible with alcohol, ether, chloroform and water.

Notes
Store in cool place. Incompatible with acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide and acids.
Specifications

Specifications

Pyrrolidine
-63°C
86°C to 88°C
99%
C4H10ClN
MFCD00005249
UN1922
Air Sensitive
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
FCLZCOCSZQNREK-UHFFFAOYSA-N
hydrogen pyrrolidine chloride
31268
71.12
Liquid
123-75-1
0.856
2°C (35°F)
Like Rotten Eggs
1.4431
100 mL
102395
14,8015
Miscible with alcohol,ether,chloroform and water.
[H+].[Cl-].C1CCNC1
107.58
CHEBI:33135
99%

Chemical Identifiers

123-75-1
107.58
FCLZCOCSZQNREK-UHFFFAOYSA-N
31268
hydrogen pyrrolidine chloride
C4H10ClN
MFCD00005249
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
CHEBI:33135
[H+].[Cl-].C1CCNC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H314-H318-H332
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H332-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRROLIDINE

EINECSNumber : 204-648-7

RTECSNumber : UX9650000

TSCA : Yes

storageNote1 : Air sensitive

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only