missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pyrromethene 556 96.0+%, TCI America™
Supplier: TCI America D4593200MG
Specifications
[[(3,5-Dimethyl-4-sulfo-1H-pyrrol-2-yl)(3,5-dimethyl-4-sulfo-2H-pyrrol-2-ylidene)methyl]methane](dif | |
C14H15BF2N2Na2O6S2 | |
200 mg | |
FBAASMARNAYERO-UHFFFAOYSA-L | |
disodium 2,2-difluoro-4,6,8,10,12-pentamethyl-1λâµ,3-diaza-2-boratricyclo[7.3.0.0³,â·]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide-5,11-disulfonate | |
14766995 | |
≥96.0% (HPLC) |
121461-69-6 | |
MFCD00467373 | |
Pyrromethene 556 | |
[Na+].[Na+].CC1=C(C(C)=C2N1[B-](F)(F)[N+]1=C(C)C(=C(C)C1=C2C)S([O-])(=O)=O)S([O-])(=O)=O | |
466.19 | |
466.19 | |
Crystalline Powder |
Chemical Identifiers
121461-69-6 | |
466.19 | |
FBAASMARNAYERO-UHFFFAOYSA-L | |
14766995 | |
[Na+].[Na+].CC1=C(C(C)=C2N1[B-](F)(F)[N+]1=C(C)C(=C(C)C1=C2C)S([O-])(=O)=O)S([O-])(=O)=O |
C14H15BF2N2Na2O6S2 | |
MFCD00467373 | |
Pyrromethene 556 | |
disodium 2,2-difluoro-4,6,8,10,12-pentamethyl-1λâµ,3-diaza-2-boratricyclo[7.3.0.0³,â·]dodeca-1(12),4,6,8,10-pentaen-1-ylium-2-uide-5,11-disulfonate |
Safety and Handling
TSCA : No