missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pyruvic Acid, 95-100%, Spectrum™ Chemical

$181.82 - $49681.59

Chemical Identifiers

CAS 127-17-3
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18613003
View Documents
Spectrum Chemical Mfg Cor
PY1105GM
5 g Amber Glass Bottle
Each for $181.82
 
18613000
View Documents
Spectrum Chemical Mfg Cor
PY110100GM
100 g Amber Glass Bottle
Each for $503.18
 
18613006
View Documents
Spectrum Chemical Mfg Cor
PY110500GM
500 g Amber Glass Bottle
Each for $1,818.74
 
18613004
View Documents
Spectrum Chemical Mfg Cor
PY1105KGBL
5 kg Amber Glass Bottle
Each for $8,416.62
 
18613001
View Documents
Spectrum Chemical Mfg Cor
PY11012KGBL
12 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18613005
View Documents
Spectrum Chemical Mfg Cor
PY11050KGBL
50 kg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

127-17-3

Specifications

127-17-3
165°C
5 g
Reagent
1
Amber Glass Bottle
95 to 100%
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.