Learn More
Pyruvic aldehyde, 35-45% w/w aq. soln, Thermo Scientific Chemicals
$53.19 - $439.66
Chemical Identifiers
CAS | 78-98-8 |
---|---|
Molecular Formula | C3H4O2 |
Molecular Weight (g/mol) | 72.06 |
MDL Number | MFCD00006960 |
InChI Key | AIJULSRZWUXGPQ-UHFFFAOYSA-N |
Synonym | methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal |
PubChem CID | 880 |
ChEBI | CHEBI:17158 |
IUPAC Name | 2-oxopropanal |
SMILES | CC(=O)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2466414
|
Thermo Scientific Chemicals
B2466414 |
25 g |
Each for $53.19
|
|
|||||
AAB2466422
|
Thermo Scientific Chemicals
B2466422 |
100 g |
Each for $123.81
|
|
|||||
AAB2466436
|
Thermo Scientific Chemicals
B2466436 |
500 g |
Each for $439.66
|
|
|||||
Description
Pyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyruvic aldehyde is used in the synthesis and cytotoxicity evaluation of N-terminal analogues of tubulysin-U an antimitotic compound. Also used in the synthesis of imidazoles as potent calcitonin gene-related CGRP antagonists.
Solubility
Soluble in DMSO, water.
Notes
Air sensitive. Store under dry inert gas. Store away from oxidizing agents, air.
Chemical Identifiers
78-98-8 | |
72.06 | |
AIJULSRZWUXGPQ-UHFFFAOYSA-N | |
880 | |
2-oxopropanal |
C3H4O2 | |
MFCD00006960 | |
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal | |
CHEBI:17158 | |
CC(=O)C=O |
Specifications
78-98-8 | |
C3H4O2 | |
CH3COCHO | |
25 g | |
Air Sensitive | |
methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal | |
AIJULSRZWUXGPQ-UHFFFAOYSA-N | |
2-oxopropanal | |
880 | |
72.06 | |
Pyruvic aldehyde |
1.2 | |
1.405 | |
MFCD00006960 | |
906750 | |
14,6081 | |
Soluble in DMSO,water. | |
CC(=O)C=O | |
72.06 | |
CHEBI:17158 | |
35 to 45% w/w aq. soln |
Safety and Handling
GHS H Statement
H301-H341-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P210-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c
H227-H302-H317-H318-H341
EINECSNumber : 201-164-8
RTECSNumber : UZ0700000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only