Learn More
Pyruvic aldehyde dimethyl acetal, 97+%, Thermo Scientific Chemicals
$93.05 - $281.23
Chemical Identifiers
CAS | 6342-56-9 |
---|---|
Molecular Formula | C5H10O3 |
Molecular Weight (g/mol) | 118.132 |
MDL Number | MFCD00008758 |
InChI Key | ULVSHNOGEVXRDR-UHFFFAOYSA-N |
Synonym | 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal |
PubChem CID | 80650 |
IUPAC Name | 1,1-dimethoxypropan-2-one |
SMILES | CC(=O)C(OC)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2498822
|
Thermo Scientific Chemicals
B2498822 |
100 g |
Each for $93.05
|
|
|||||
AAB2498836
|
Thermo Scientific Chemicals
B2498836 |
500 g |
Each for $281.23
|
|
|||||
Description
Pyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyruvic aldehyde dimethyl acetal was used in the synthesis of methylglyoxal via hydrolysis in the presence of H2SO4. It was used to investigate the effects of methylglyoxal-mediated glycation on the structure, thermal stability and enzyme activity of yeast enolase.
Solubility
Soluble in water. 350 g/L at 20°C.
Notes
Do not store together with acids. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
6342-56-9 | |
118.132 | |
ULVSHNOGEVXRDR-UHFFFAOYSA-N | |
80650 | |
CC(=O)C(OC)OC |
C5H10O3 | |
MFCD00008758 | |
1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal | |
1,1-dimethoxypropan-2-one |
Specifications
6342-56-9 | |
0.998 | |
34°C (93°F) | |
1.398 | |
100 g | |
1560557 | |
Soluble in water. 350g/L at 20°C. | |
CC(=O)C(OC)OC | |
118.132 | |
118.13 | |
Pyruvic aldehyde dimethyl acetal |
-57°C | |
133°C to 135°C | |
C5H10O3 | |
MFCD00008758 | |
UN1993 | |
1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal | |
ULVSHNOGEVXRDR-UHFFFAOYSA-N | |
1,1-dimethoxypropan-2-one | |
80650 | |
≥97% |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 228-735-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only