Learn More
MP Biomedicals™ pZerve™ Cryopreservation Solution
pZerve is a ready-to-use cryopreservation solution that does not contain dimethyl sulfoxide (DMSO), fetal bovine serum or other animal protein.
Supplier: MP Biomedicals™ 0920303M2
Description
pZerve is a ready-to-use, sterile, cryopreservation solution that does not contain dimethyl sulfoxide (DMSO), fetal bovine serum or other animal protein. pZerve does not require any dilution or further processing and can be used for cells cultured in serum free medium or with serum containing medium.
pZerve has been successfully used with the following cell types including, but not limited to:
- MastCells
- Dendritic Cells
- Mesenchymal Cells
- LNCaP - Metastatic Prostate Adenocarcinoma, Human
- LS 180 - Colon, Adenocarcinoma, Human
- 293 - Transformed Primary Embryonal Kidney, Human
Specifications
Stored at 2 to 8°C | |
Cryopreservation Media | |
Cryopreservation |
Liquid | |
Sterile | |
60 mL |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.