missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ pZerve Cryopreservation Solution
A cryopreservation solution that does not contain dimethyl sulfoxide (DMSO) and serum.
$548.10 - $548.10
Specifications
Content And Storage | Store at 2° to 8°C |
---|---|
Description | pZerve Cryopreservation Media |
Form | Solution |
Product Type | Cryopreservation Media |
For Use With (Application) | Cryopreservation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
MP92030346
|
MP Biomedicals™
092030346 |
20Â mL |
Each for $548.10
|
|
||||||
Description
pZerve has been successfully used with a variety of cell types including mast cells, dendritic cells, and mesenchymal cells. It can be used for cells cultured in serum-free medium or with serum-containing medium.
Specifications
Store at 2° to 8°C | |
Solution | |
Cryopreservation |
pZerve Cryopreservation Media | |
Cryopreservation Media |