Qorpak™ Amber Borosilicate Vials, Vacuum and Ionized
Individually cleaned of contaminants and sealed
Supplier:  Qorpak™ GLC06031
 View more versions of this product
Catalog No. 02-991-857
Explore available promotions
Description
Description
  • Air inside the bottle is odor-free, tasteless and does not react with product
  • Separate washing not required, thereby saving time
  • Final finished product kept clean and dust-free
  • With green Thermoset F217 and PTFE caps attached
Specifications
Specifications
F217/PTFE
Amber
Borosilicate Glass
Vacuum and ionized
Screw
Thermoset
70 mm
0.81 in.
20.7 mm
15 mL
2.75 in.
4 dr.
Screw Cap
144/Cs.