Qorpak™ CurTec™ Wide Neck Drums
Provides high level of product protection and added value through quality, functionality, ease of use, and design. Qorpack™ CurTec™ Wide Neck Drums are used for shipping and stocking valuable fine chemicals.
Supplier: Qorpak™ 248723
Description
- Features red polypropylene unlined lid
- Wide neck for easy filling of contents
Specifications
HACCP, ISO | |
121.7 oz. | |
White | |
7.7 in. | |
6.5 in. | |
21 lb. |
Cap | |
3.6 L | |
Ideal for pharmaceuticals, fine chemicals, food ingredients and hazardous waste | |
198 mm | |
165 mm | |
9.5 Kg |