Qsonica Cup Horns
Ideal for sterile or pathogenic sample processing
Supplier:  Qsonica 431C2
Catalog No. 15-338-316
Explore available promotions
Description
Description
  • Multiple samples can be processed
  • Offers indirect sonication and function as high intensity ultrasonic water baths
  • Avoids cross contamination
  • Horn is mounted in acrylic cup
  • Available in two sizes: 3 in. or 5.5 in.
  • Replacement microtube racks sold separately (Cat. Nos. 15-338-404 and -405)
Specifications
Specifications
Cup Horn
Ideal for sterile or pathogenic sample processing
20 tubes
5.5 in.
Q700 Sonicator™ without probe
Sample rack (part# 440)