missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Quant-iT™ dsDNA Assay Kits, high sensitivity (HS) and broad range (BR) DFS Item

For highly selective quantitation of double-stranded DNA over RNA, the Quant-iT dsDNA Assay Kits, high sensitivity and broad range, produce fluorescence signals that are linear in the ranges of 0.2-100 ng and 2-1000 ng of DNA, respectively.

Supplier:  Invitrogen™ Q33120

 View more versions of this product

Catalog No. Q33120


Explore available promotions

Description

Description

Perform dsDNA quantification easily and quickly with Quant-iT dsDNA assay kits. Both the Quant-iT High-Sensitivity dsDNA Assay Kit and the Quant-iT Broad-Range dsDNA Assay Kit provide concentrated assay reagent, dilution buffer, and pre-diluted DNA standards. These DNA assay kits are highly selective for double-stranded DNA over RNA, and, in the ranges of 0.2–100 ng DNA (for the HS dsDNA Assay Kit) and 4–1000 ng DNA (for the BR dsDNA Assay Kit), the fluorescence signal is linear.

The Quant-iT dsDNA High-Sensitivity Assay Kit and the Quant-iT dsDNA Broad-Range Assay Kit make DNA quantification easy and accurate. The kits provide concentrated assay reagent, dilution buffer, and prediluted DNA standards. Simply dilute the reagent 1:200, load 200 μL into the wells of a microplate, add 1–20 μL sample, mix, then read the fluorescence.

The assays are highly selective for double-stranded DNA over RNA, and, in the range of 0.2–100 ng for the HS assay or 4–1000 ng for the BR assay, the fluorescence signal is linear with DNA. The assays are performed at room temperature, and the signal is stable for three hours. Common contaminants such as salts, solvents, detergents, and protein are well tolerated in these assays.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

dsDNA Quantitation, high sensitivity
Microplate Reader
0.2 to 100 ng
1 kit
Fluorescence
1000 Assays (200 μL assay volume)
Quant-iT
Room Temperature
Videos
SDS
Documents

Documents

Promotions

Promotions