missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ QuantaRed™ Enhanced Chemifluorescent HRP Substrate Kit

Enable sensitivity comparable to enhanced chemiluminescence (ECL) using the most sensitive fluorescent ELISA substrate for peroxidase (HRP).

Supplier:  Thermo Scientific™ 15159

Catalog No. PI15159


Explore available promotions

Description

Description

Thermo Scientific™ QuantaRed Enhanced Chemifluorescent HRP Substrate is the most sensitive fluorescent ELISA substrate, providing sensitivity comparable to enhanced chemiluminescence.

QuantaRed Enhanced Chemifluorescent HRP Substrate enables high-yield and sensitive fluorescent and colorimetric detection of peroxidase activity in the ELISA format. QuantaRed Substrate uses a proprietary technology to increase fluorescent yield and sensitivity of the well-documented ADHP (10-acetyl-3,7-dihydroxyphenoxazine) chemifluorescence reaction. ADHP is a non-fluorescent compound that reacts with horseradish peroxidase (HRP) to produce resorufin, a soluble, highly fluorescent reaction product (excitation/emission approx. 570/585nm). The long wavelength emission of the resorufin minimizes interference from the low wavelength (blue and green) autofluorescence, making it an ideal substrate for quantitating target molecules in biological samples. In addition, the colored reactopm product of QuantaRed Substrate can be measured spectrophotmetrically at 570nm.

Highlights:

  • Sensitive – low picogram to femtogram detection

  • Red-shifted – less interference from autofluorescence generated by biological samples

  • Convenient – compatible with existing filter sets, no need to purchase special filter sets

  • Complete – easy to use; no buffers to prepare or reagent powders to dissolve

  • Flexible – effective stop solution result in stable signal for at least 4 hours

  • Versatile – produces strong signal for easy fluorometric or colorimetric measurement

Note:

Equilibrate all reagents to room temperature before use. Temperature can affect the reaction kinetics.

Specifications

Specifications

ELISA Substrate
Fluorescence
Chemical Substrate
Sufficient For: 960 wells (10 x 96-well plates) at 100 μL working solution per well
• QuantaRed ADHP Concentrate, 1 mL
• QuantaRed Enhancer Solution, 50 mL
• QuantaRed Stable Peroxide Solution, 50 mL
• QuantaRed Stop Solution, 10 mL

Store at 4°C.
110 ml
QuantaRed™
HRP (Horseradish Peroxidase)
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.