missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Applied Biosystems™ QuantStudio 3 qPCR System (0.2mL) and TaqMan Master Mix Bundle
Applied Biosystems™ QuantStudio qPCR System (0.2mL) and Master Mix Bundle is for researchers performing gene expression who need an intuitive and easy-to-use qPCR solution. Offers excellent specificity with the speed and ease-of-use of Cloud connectivity
Supplier: Applied Biosystems™ A39941
Includes:
- QuantStudio 3 qPCR System, 96-well, 0.2mL, Laptop
- SuperScript IV VILO Master Mix with ezDNase (50rxn)
- TaqMan Fast Advanced Master Mix (5 x 5mL)
- MicroAmp Fast Optical 96-Well Reaction Plate, 0.2mL (10 plates)
- MicroAmp Optical Adhesive Film (100 covers)
Description
QuantStudio and Real-Time PCR Systems provide a modern usability experience similar to mobile devices, allowing users to connect to data easier than ever before.
- Access, analyze, and share data anytime, anywhere
- Support remote monitoring of your runs
- Quick analysis of sophisticated data sets
- Secure storage of results
- Safe sharing of results online with colleagues across campus and around the world with web browser-based software
Warranty
1 year
Specifications
Non-interchangeable | |
0.4°C | |
100 to 240 V | |
16 in. | |
40 cm | |
1 Year | |
50 cm | |
qPCR | |
50 cm | |
960 W | |
26 kg (57 lb.) |
0.25°C | |
96-well; 10 to 100μL | |
QuantStudio Real Time PCR System (qPCR) | |
20 in. | |
27 cm | |
10.5 in. | |
0°C to 100°C | |
±3.66 °C/s | |
Touchscreen | |
6.5°C/s |