Applied Biosystems™ QuantStudio™ Absolute Q™ MAP16 Plate Kit and Master Mix
A key component of the QuantStudio Absolute Q Digital PCR System powered by the patented microfluidic array plate (MAP) technology, enabling consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV).
Supplier: Applied Biosystems™ A53301
Description
- A key component of the QuantStudio Absolute Q Digital PCR System
- System is powered by the patented microfluidic array plate (MAP) technology that enables consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV)
- QuantStudio Absolute Q MAP16 consumable uses microinjection-molded plates to overcome common challenges of inconsistency and high dead volume
- Benefits of this approach include precise microchamber volume, consistent sample distribution, and greater than 98% of loaded volume analyzed
- Includes QuantStudio Absolute Q DNA Master Mix optimized for use with the QuantStudio Absolute Q Digital PCR System and Applied Biosystems digital PCR (dPCR) assays
- 5X formulation enables analysis of higher sample volumes and delivers accurate quantification of DNA targets without using a standard curve
- Unparalleled precision and accuracy of the Absolute Q system provides confidence in measurements across replicates and experiments
Specifications
Includes Barcode | |
Digital PCR | |
12 MAP16 Plates, Gaskets, Isolation Buffer, and Absolute Q™ DNA Digital PCR Master Mix | |
12 plates |
For Research Use Only. Not for use in diagnostic procedures