missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Qubit™ Endotoxin Detection Assay Kit

The Invitrogen Qubit Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on the Qubit Flex Fluorometer that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples.

Supplier:  Invitrogen™ Q32891

Catalog No. Q32891


Explore available promotions

Description

Description

The Invitrogen Qubit Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on the Qubit Flex Fluorometer that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples. Amebocyte lysates are widely used as a simple and sensitive assay for the detection of endotoxin lipopolysaccharide (LPS), the membrane component of gram-negative bacteria.

Features of the Qubit Endotoxin Detection Assay Kit include:
Highly sensitive with a broad range—detect as little as 0.01 EU/mL to 10.0 EU/mL
Specific—no interference from ß-glucans and suitable for wide range of samples, including protein, vaccine, plasmid, DNA, and RNA
Easy to use—when paired with the Qubit Flex Fluorometer, calculations are performed automatically, reducing the potential for error

Endotoxin levels in the samples are accurately determined using the included endotoxin standard of known concentration that is derived from E. coli strain O111: B4. Determining endotoxin levels is important to assess the efficiency of endotoxin removal methods and prevent endotoxic shock, inflammation, and/or sepsis in tissue culture cells and animals injected with endotoxin-contaminated proteins.

When endotoxin encounters the amebocyte lysate, a series of enzymatic reactions results in the activation of Factor C, Factor B, and pro-clotting enzyme. The activated enzyme catalyzes a cleavage event in the substrate to produce a strong fluorescent signal. After stopping the reaction, the resulting signal is measured on the Qubit Flex Fluorometer. The correlation between fluorescent signal is proportional to the endotoxin concentration in the sample.

Notes
• Designed for use with the Qubit Flex Fluorometer (Cat. No. Q33327) and not currently for use on Qubit 4 Fluorometers
• Optimized for use with Qubit Flex Pyrogen Free Assay Tube Strips (Cat. No. Q32893)
• Use the Qubit Flex System Verification Assay Kit (Cat. No. Q33254) prior to first assay run

Specifications

Specifications

Endotoxin
Kit
Qubit Flex Fluorometer
Fluorescence
Protein, Antibody, Plasmid, DNA, RNA
80 Reactions
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.