Invitrogen™ Qubit™ Flex Assay Tube Strips
Strips of eight tubes designed for the Qubit™ Flex.
Supplier:  Invitrogen™ Q33252
Catalog No. Q33252
Explore available promotions
Includes: 125 tube strips
Description
Description
Qubit™ Flex Assay Tube Strips are 200-μL thin-walled polypropylene tubes in a strip of eight tubes that are for use with the Qubit™ Flex Fluorometer.
Specifications
Specifications
Room Temperature
Qubit™
200 μL
Assay Tube Strip
125 tube strips
Qubit Flex Fluorometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.