Invitrogen™ Qubit™ Flex Pyrogen-Free Tube Strips
Invitrogen Qubit Flex Pyrogen Free Tube Strips are clear 8 x 200-μL thin-walled tubes with attached flat caps.
Supplier: Invitrogen™ Q32893
Description
Invitrogen Qubit Flex Pyrogen Free Tube Strips are clear 8 x 200-μL thin-walled tubes with attached flat caps designed for use with the Qubit Flex Fluorometer (Cat. No. Q33327) and the Qubit Endotoxin Detection Assay Kit (Cat. No. Q32891).
Notes
• For DNA, RNA, and protein quantification use Qubit Flex Assay Tube Strips (Cat. No. Q33252).
Specifications
Assay Tube Strip | |
120 strips | |
Qubit Flex Fluorometer |
For Research Use Only. Not for use in diagnostic procedures.