missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quercetin hydrate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 174070100

 View more versions of this product

Catalog No. AC174070100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Quercetin hydrate
>300°C
Authentic
Glass bottle
MFCD03847906
18, 242
quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
REFJWTPEDVJJIY-UHFFFAOYSA-N
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
16212154
95%
849061-97-8
Yellow
94% min. (HPLC)
C15H10O7
10 g
14, 8034
Solubility in water: practically insoluble. Other solubilities: soluble in ethanol,acetone,pyridine,,glacial acetic acid
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1
302.24
302.24
Crystalline Powder

Chemical Identifiers

849061-97-8
302.24
REFJWTPEDVJJIY-UHFFFAOYSA-N
16212154
OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC(O)=C(O)C=C2)=C1
C15H10O7
MFCD03847906
quercetin hydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, quercetinhydrate, quercetine sophoretin /, regid_for_cid_747934, regid_for_cid_16212154, 3,3;,4;,5,7-pentahydroxyflavon, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxychromen-4-one, hydrate
2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash face,hands and any exposed skin thoroughly after handling.

GHS Signal Word: Danger

RTECSNumber : LK8950000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only