Innovatek QuickCheck Urine Drug Screen Tests
Innovatek QuickCheck Urine Drug Screen Tests is able to detect twelve commonly abused drugs
$302.58 - $345.79
Specifications
Certifications/Compliance | US SAMSHA, DOT |
---|---|
Detectable Analytes | Amphetamine, Barbiturates, Benzodiazepine, Buprenorphine, Cocaine, Creatinine, Methadone Metabolite, Opiate, Oxycodone, Marijuana, Methamphetamine, Phencyclidine |
Disposable | Single-use |
For Use With (Application) | Drug Testing |
Test Time | 5 min. |
Description
- Accurate and sensitive test procedure
- Built-in test controls
- Clear, easy-to-read, precise test results within 5 minutes
- Meets US SAMSHA and DOT requirements
- Creatinine test (CRE) included
- Temperature indicator on drug screen cups
Specifications
US SAMSHA, DOT | |
Single-use | |
5 min. |
Amphetamine, Barbiturates, Benzodiazepine, Buprenorphine, Cocaine, Creatinine, Methadone Metabolite, Opiate, Oxycodone, Marijuana, Methamphetamine, Phencyclidine | |
Drug Testing |