Innovatek QuickStep™ Plus Pregnancy Cassette Tests
Innovatek QuickStep™ Plus Pregnancy Cassette Tests provide results within 5 minutes
Supplier:  Innovatek 422033
 View more versions of this product
Catalog No. 29-801-09
Explore available promotions
Description
Description
  • Patient ID can be labeled on each cassette
  • For urine, or urine/serum testing
  • Individually foil wrapped, includes pipette and desiccant
  • Pipette sample into well and read in 5 minutes
Specifications
Specifications
Urine
Pregnancy Testing
20 Tests
≈5 min.