missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quidel QuickVue™ Dipstick Strep A Test

Detects Group A Streptococcal antigen directly from patient throat swab specimens or confirms presumptive Group A Streptococcal colonies recovered from culture. Used as an aid in the diagnosis of Group A Streptococcal infection. Lateral flow immunoassay, using Quidel's patented assay device.

Supplier:  Quidel Q20108IN

 View more versions of this product

Catalog No. 23043051


Explore available promotions

Description

Description

  • CLIA Complexity: Waived
  • CPT Codes: 87880/87880QW
  • Sensitivity: 92%
  • Specificity: 98%
  • Overall Accuracy: 95%
  • Time to Results: Read at five minutes
  • Specimen Type: Throat Swab
  • Internal Procedure Controls: Positive and negative
  • External Controls: Positive and negative liquid

Patent No: 5770460 (June 2015)

Specifications

Specifications

Room temperature for up to 24 months after date of manufacture.
Waived
Throat Swab
50 Tests/Kit
87880, 87880QW
Streptococcal Antigen (Group A)
Dipstick Streptococcus A Test
5 min.
Videos
SDS
Documents

Documents

Promotions

Promotions