missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quidel QuickVue™ In-Line™ One-Step Strep A Test
Unique patented in-line extraction method reduces hands-on time and provides quick results
Supplier: Quidel Q00343IN
Includes: Reaction units, extraction reagents, throat swabs, package insert, and positive and negative controls
Description
- CLIA Complexity: Waived
- CPT Codes: 87880/87880QW
- One-step lateral flow immunoassay for the detection of Group A streptococcal antigen from throat swab specimens
- Results within 5 minutes
- Sensitivity: 92%
- Specificity: 99%
- Overall accuracy: 96%
- Ideal for exam room environments
- Built-in procedural controls confirm proper test performance
- Two-color read-out makes results easy to interpret
- For in vitro diagnostic use
Specifications
| 87880, 87880QW | |
| Streptococcal Antigen (Group A) | |
| Streptococcus A Test | |
| 25 Tests |
| Waived | |
| Throat Swab | |
| Reaction units, Extraction reagents, Throat swabs, Package insert and Positive and Negative control | |
| 5 min. |