Quidel QuickVue™ H. pylori Test
Rapid immunoassay for the simple, accurate and qualitative detection of IgG antibodies specific to H. pylori
$141.16 - $423.47
Specifications
Content And Storage Room Temperature
Clia Complexity Waived, Non-waived
Detectable Analytes H. pylori IgG
Type H. pylori Test Kit
Includes Testing cassettes, positive and negative control, transfer pipets and capillary tubes, package insert and procedure card, positive and negative external control
View More Specs
Includes: Each complete kit contains testing cassettes, positive and negative controls, transfer pipets and capillary tubes, package insert and procedure card
Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
120116
View Documents
Quidel
Q0W009
10 Tests
Pack of 10 Each for $141.16
120117
View Documents
Quidel
Q0W010
30 Tests
Pack of 30 Each for $423.47
Description
Description
  • CLIA Complexity: Waived (whole blood); Non-waived (serum and plasma)
  • CPT Codes: 86318 (serum/plasma), 86318QW (whole blood)
  • Lateral-flow immuno-chromatographic assay
  • Test results in five minutes; 92% overall agreement with EIA
  • Tests serum and plasma
  • Two-color endpoint for easy-to-read and interpret results
  • Simple one-step procedure from only two hanging drops of blood; serum or plasma can also be used
  • For in vitro diagnostic use
Specifications
Specifications
Room Temperature
H. pylori IgG
Testing cassettes, positive and negative control, transfer pipets and capillary tubes, package insert and procedure card, positive and negative external control
Waived, Non-waived
H. pylori Test Kit
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'