missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quidel QuickVue™ hCG Combo Test Kit

Uses a monoclonal antibody specific to the beta subunit of hCG in a single-step technology

Supplier:  Quidel Q20110IN

Catalog No. 23043053


Explore available promotions

Includes: Individually wrapped test cassettes, disposable pipets, procedure card, and package insert

Description

Description

  • CPT Codes: 81025 (urine), 84703 (serum)
  • CLIA Complexity: Waived (urine), Non-waived (serum)
  • For the qualitative detection of human chorionic gonadotropin (hCG) in serum or urine for the early detection of pregnancy
  • Sensitivity: >99%
  • Specificity: >99%
  • Agreement: >99%
  • Results in three minutes for urine, five minutes for serum
  • Positive results: pink-to-red Test (T) line along with a blue procedural Control (C) line; negative results: only a blue procedural Control line. Specimens containing as low as 25mIU/mL (calibrated against the WHO 4th IS 75/589) hCG will yield positive results.
  • For in vitro diagnostic use

Specifications

Specifications

>99%
Waived, Non-waived
Serum, Urine
Individually wrapped test cassettes, disposable pipets, procedure card and package insert
3 Drops
81025, 84703
hCG
hCG Combo Test Kit
50 Tests
3 min. for urine, 5 min. for serum
Videos
SDS
Documents

Documents

Promotions

Promotions