missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quidel QuickVue+™ Strep A Liquid Control
For use with Quidel QuickVue+ Strep A Tests
Supplier: Quidel 00354
Description
- Contains one positive and one negative control
Specifications
Room Temperature | |
Streptococcus A Liquid Control | |
65 Tests |
Streptococcal Antigen (Group A) | |
Positive and negative control |