missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quidel QuickVue+™ Strep A Test
![Edge](/content/dam/fishersci/glyphs/edge.png)
Built-in controls satisfy CLIA requirements for daily QC
Supplier: Quidel Q20122IN
Includes: Positive and negative external controls
Description
- CLIA Complexity: Non-waived
- Sensitivity: 95%
- Specificity: 98%
- Lateral flow immunoassay for the detection of group A streptococcal antigen directly from throat swabs and β-hemolytic colonies on blood agar plates
- Clearly distinguishable +/- endpoint and two reagent extraction make testing simple
- Results in five minutes
- Shelf life: 18 months from the date of manufacture
Specifications
Non-waived | |
Throat swab and beta-hemolytic colonies on Blood Agar plates | |
Positive and negative external control | |
5 min. |
Streptococcal Antigen (Group A) | |
Streptococcus A Test | |
25 Tests |