missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals Quinaldine Red, dye content, 95%

Supplier:  Thermo Scientific Chemicals H5572906

 View more versions of this product

Catalog No. AAH5572906


Explore available promotions

Description

Description

Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quinaldine Red is a colorimetric phosphate detection reagent which is known to be a cationic fluorescent probe for proteins. It is also a pH indicator with a transition interval from 1.4 - 3.2 (colorless - rose).

Solubility
Partly soluble in water, alcohol, and ethanol.

Notes
Store at room temperature. Avoid strong oxidizing agents.
Specifications

Specifications

Quinaldine Red
117-92-0
C21H23IN2
5 g
Light sensitive
quinaldine red, eastman 1361, 2-p-dimethylaminostyryl quinoline ethiodide, 1-ethyl-2-p-dimethylaminostyrylquinoline iodide, 2-4-dimethylaminostyryl-1-ethylquinolinium iodide, 2-4-dimethylamino styryl-1-ethylquinolinium iodide, 2--4-dimethylamino styryl-1-ethylquinolinium iodide, chinaldinrot, alpha-p-dimethylaminophenylethylene quinolineethiodide, quinolinium, 2-2-4-dimethylamino phenyl ethenyl-1-ethyl-, iodide 1:1
JOLANDVPGMEGLK-UHFFFAOYSA-M
4-[(E)-2-(1-ethylquinolin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide
5702759
95%
Dye content
Black to Green
MFCD00011968
3798055
14,8049
Partly soluble in water,alcohol,and ethanol.
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
430.33
430.33
Powder

Chemical Identifiers

117-92-0
430.33
JOLANDVPGMEGLK-UHFFFAOYSA-M
5702759
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C
C21H23IN2
MFCD00011968
quinaldine red, eastman 1361, 2-p-dimethylaminostyryl quinoline ethiodide, 1-ethyl-2-p-dimethylaminostyrylquinoline iodide, 2-4-dimethylaminostyryl-1-ethylquinolinium iodide, 2-4-dimethylamino styryl-1-ethylquinolinium iodide, 2--4-dimethylamino styryl-1-ethylquinolinium iodide, chinaldinrot, alpha-p-dimethylaminophenylethylene quinolineethiodide, quinolinium, 2-2-4-dimethylamino phenyl ethenyl-1-ethyl-, iodide 1:1
4-[(E)-2-(1-ethylquinolin-1-ium-2-yl)ethenyl]-N,N-dimethylaniline;iodide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-221-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only