missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinaldine Red 95.0+%, TCI America™
$34.84 - $248.86
Chemical Identifiers
CAS | 117-92-0 |
---|---|
Molecular Formula | C21H23IN2 |
Molecular Weight (g/mol) | 430.33 |
MDL Number | MFCD00011968 |
InChI Key | JOLANDVPGMEGLK-UHFFFAOYSA-M |
PubChem CID | 5702759 |
IUPAC Name | 2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide |
SMILES | [I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C |
Chemical Identifiers
117-92-0 | |
430.33 | |
JOLANDVPGMEGLK-UHFFFAOYSA-M | |
2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide |
C21H23IN2 | |
MFCD00011968 | |
5702759 | |
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C |
Specifications
117-92-0 | |
MFCD00011968 | |
[I-].CC[N+]1=C2C=CC=CC2=CC=C1C=CC1=CC=C(C=C1)N(C)C | |
430.33 | |
430.33 | |
Black/Green | |
Crystal/Powder |
C21H23IN2 | |
JOLANDVPGMEGLK-UHFFFAOYSA-M | |
2-{2-[4-(dimethylamino)phenyl]ethenyl}-1-ethylquinolin-1-ium iodide | |
5702759 | |
≥95.0% (T) | |
1 g | |
Quinaldine Red |
Safety and Handling
TSCA : Yes