missing translation for 'onlineSavingsMsg'
Learn More

Quinhydrone, 98%

Quinhydrone, 98%, Quantity: 100g, Packaging: Glass bottle, Green, Melting Point: 168 deg.C to 172 deg.C, Molecular Weight: 218.21, Percent Purity: 98%, Assay Percent Range: 48.0 to 52% Hydroquinone (HPLC), CAS: 106-34-3, ChEBI: CHEBI:26491, Density: 1.4000g/mL | CAS: 106-34-3 | C12H10O4 | 218.21 g/mol

$85.43 - $1008.27

Chemical Identifiers

CAS 106-34-3
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.21
MDL Number MFCD00010310
InChI Key BDJXVNRFAQSMAA-UHFFFAOYSA-N
Synonym quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
PubChem CID 7801
ChEBI CHEBI:26491
IUPAC Name benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
SMILES OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC418770250
SDS View Documents
Thermo Scientific Chemicals
418770250
25 g Glass bottle
Each for $85.43
Only null left
 
AC418771000
SDS View Documents
Thermo Scientific Chemicals
418771000
100 g Glass bottle
Each for $200.05
Only null left
 
AC418775000
SDS View Documents
Thermo Scientific Chemicals
418775000
500 g Glass bottle
Each for $1,008.27
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

106-34-3
218.21
BDJXVNRFAQSMAA-UHFFFAOYSA-N
7801
benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
C12H10O4
MFCD00010310
quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
CHEBI:26491
OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1

Specifications

106-34-3
Green
Authentic
Glass bottle
MFCD00010310
1.4
quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
BDJXVNRFAQSMAA-UHFFFAOYSA-N
benzene-1,4-diol;cyclohexa-2,5-diene-1,4-dione
7801
218.21
Crystalline Powder
168°C to 172°C
1.4000g/mL
48.0 to 52% Hydroquinone (HPLC)
C12H10O4
25 g
15, 8174
Solubility in water: 4g/L (20°C). Other solubilities: soluble in hot water,ammonia,alcohol,ether,insoluble in petroleum ether
OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1
218.21
CHEBI:26491
98%
Quinhydrone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Very toxic to aquatic life.
Toxic if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN

GHS Signal Word: Danger

EINECSNumber : 203-387-6

RTECSNumber : VA4550000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only