missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Quinidine Sulfate Dihydrate 98.0+%, TCI America™

Supplier:  TCI America Q00105G

 View more versions of this product

Catalog No. Q00105G


Explore available promotions

Description

Description

Specifications

Specifications

Quinidine Sulfate Dihydrate
212°C
C40H54N4O10S
5 g
bis s-6-methoxyquinolin-4-yl 1s,2s,4s,5r-5-vinylquinuclidin-2-yl methanol sulfate dihydrate
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.OS(=O)(=O)O
782.95
746.92
Crystalline Powder
6591-63-5
White-Yellow
MFCD00135926
2811
ZHNFLHYOFXQIOW-CMZPEELOSA-N
(S)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate
132274356
≥98.0% (T)

Chemical Identifiers

6591-63-5
782.95
ZHNFLHYOFXQIOW-CMZPEELOSA-N
132274356
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CCN3CC4C=C)O.O.O.OS(=O)(=O)O
C40H54N4O10S
MFCD00135926
bis s-6-methoxyquinolin-4-yl 1s,2s,4s,5r-5-vinylquinuclidin-2-yl methanol sulfate dihydrate
(S)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;sulfuric acid;dihydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0324

RTECSNumber : VA5605000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions