Learn More
Quinoline-2-carbonitrile, 97%, Thermo Scientific Chemicals
$141.58 - $480.55
Chemical Identifiers
CAS | 1436-43-7 |
---|---|
Molecular Formula | C10H6N2 |
Molecular Weight (g/mol) | 154.17 |
MDL Number | MFCD00134341 |
InChI Key | WDXARTMCIRVMAE-UHFFFAOYSA-N |
Synonym | 2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0 |
PubChem CID | 74031 |
IUPAC Name | quinoline-2-carbonitrile |
SMILES | N#CC1=CC=C2C=CC=CC2=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1942703
|
Thermo Scientific Chemicals
L1942703 |
1 g |
Each for $141.58
|
|
|||||
AAL1942706
|
Thermo Scientific Chemicals
L1942706 |
5 g |
Each for $480.55
|
|
|||||
Description
Quinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsQuinoline-2-carbonitrile is used to produce quinoline-2-carboxylic acid amide. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Insoluble in water.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Chemical Identifiers
1436-43-7 | |
154.17 | |
WDXARTMCIRVMAE-UHFFFAOYSA-N | |
74031 | |
N#CC1=CC=C2C=CC=CC2=N1 |
C10H6N2 | |
MFCD00134341 | |
2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0 | |
quinoline-2-carbonitrile |
Specifications
1436-43-7 | |
160°C (23 mmHg) | |
C10H6N2 | |
1 g | |
2-quinolinecarbonitrile, quinaldonitrile, 2-cyanoquinoline, quinolinecarbonitrile, cyanoquinoline, 2-cyano-quinoline, quinoline-2-carbinitrile, acmc-1bzd0 | |
WDXARTMCIRVMAE-UHFFFAOYSA-N | |
quinoline-2-carbonitrile | |
74031 | |
97% |
93°C to 96°C | |
Odorless | |
MFCD00134341 | |
115213 | |
Insoluble in water. | |
N#CC1=CC=C2C=CC=CC2=N1 | |
154.17 | |
154.17 | |
Quinoline-2-carbonitrile |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 215-865-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only