missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Quinoline-8-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals
$134.22 - $474.81
Chemical Identifiers
CAS | 190788-62-6 |
---|---|
Molecular Formula | C15H18BNO2 |
Molecular Weight (g/mol) | 255.124 |
MDL Number | MFCD05663894 |
InChI Key | RNTGVJBKPQOGPS-UHFFFAOYSA-N |
Synonym | quinoline-8-boronic acid pinacol ester, 8-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 8-quinolineboronic acid pinacol ester, 8-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, amtb187, quinoline-8-boronic acid, pinacol ester |
PubChem CID | 17750280 |
IUPAC Name | 8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C=CC=N3 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5169803
|
Thermo Scientific Chemicals
H5169803 |
1 g |
Each for $134.22
|
|
|||||
AAH5169806
|
Thermo Scientific Chemicals
H5169806 |
5 g |
Each for $474.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
190788-62-6 | |
255.124 | |
RNTGVJBKPQOGPS-UHFFFAOYSA-N | |
17750280 | |
B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C=CC=N3 |
C15H18BNO2 | |
MFCD05663894 | |
quinoline-8-boronic acid pinacol ester, 8-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 8-quinolineboronic acid pinacol ester, 8-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, amtb187, quinoline-8-boronic acid, pinacol ester | |
8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline |
Specifications
190788-62-6 | |
MFCD05663894 | |
quinoline-8-boronic acid pinacol ester, 8-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, 8-quinolineboronic acid pinacol ester, 8-tetramethyl-1,3,2-dioxaborolan-2-yl quinoline, amtb187, quinoline-8-boronic acid, pinacol ester | |
B1(OC(C(O1)(C)C)(C)C)C2=C3C(=CC=C2)C=CC=N3 | |
255.124 | |
255.12 | |
Quinoline-8-boronic acid pinacol ester |
C15H18BNO2 | |
1 g | |
RNTGVJBKPQOGPS-UHFFFAOYSA-N | |
8-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)quinoline | |
17750280 | |
95% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only