Stoelting™ Quintessential Stereotaxic Injector (QSI)
Program injection parameters (volume, rate and syringe specifications) using the easy-to-use touch screen control box.
Supplier:  Stoelting™ 53312
 View more versions of this product
Catalog No. 10-000-005
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
  • Control one or two independent injections
  • Automatic infusion or withdrawal of nL-μL volumes
  • Accommodates Hamilton syringes sized 0.5 to 250 uL
  • Available as a single or dual injector
  • If purchased as a single injector, add a second injector at any time.
  • Preprogrammed syringe specifications
  • Out-of-range warnings
Specifications
Specifications
Direct injection into the central nervous system using Hamilton microsyringes
Stereotaxic Instrument
Mouse, Rat
2 Injector h=Heads, 1 Touchscreen Controller