Learn More
(R)-(+)-1-(1-Naphthyl)ethylamine, 99%
CAS: 3886-70-2 | C12H13N | 171.243 g/mol
$270.10 - $884.42
Chemical Identifiers
| CAS | 3886-70-2 |
|---|---|
| Molecular Formula | C12H13N |
| Molecular Weight (g/mol) | 171.243 |
| MDL Number | MFCD00064114 |
| InChI Key | RTCUCQWIICFPOD-SECBINFHSA-N |
| Synonym | r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine |
| PubChem CID | 2724264 |
| IUPAC Name | (1R)-1-naphthalen-1-ylethanamine |
| SMILES | CC(C1=CC=CC2=CC=CC=C21)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1691806
|
Thermo Scientific Chemicals
A1691806 |
5 g |
Each for $270.10
|
|
|||||
|
AAA1691814
|
Thermo Scientific Chemicals
A1691814 |
25 g |
Each for $884.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3886-70-2 | |
| 171.243 | |
| RTCUCQWIICFPOD-SECBINFHSA-N | |
| 2724264 | |
| CC(C1=CC=CC2=CC=CC=C21)N |
| C12H13N | |
| MFCD00064114 | |
| r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine | |
| (1R)-1-naphthalen-1-ylethanamine |
Specifications
| 3886-70-2 | |
| 152°C to 153°C (11 mmHg) | |
| Amine-like | |
| 1.623 | |
| 5 g | |
| 2208025 | |
| r-+-1-1-naphthyl ethylamine, r-1-naphthalen-1-yl ethanamine, 1r-1-naphthalen-1-yl ethan-1-amine, r-1-1-naphthyl ethylamine, 1r-1-naphthalen-1-ylethanamine, 1r-1-naphthylethylamine, r-+-alpha-1-naphthyl ethylamine, 1r-1-1-naphthyl ethylamine, r-+-a-1-naphthyl ethylamine, 1r-1-naphthalen-1-yl ethanamine | |
| CC(C1=CC=CC2=CC=CC=C21)N | |
| 171.243 | |
| 171.24 | |
| (R)-(+)-1-(1-Naphthyl)ethylamine |
| 1.06 | |
| >110°C (230°F) | |
| C12H13N | |
| MFCD00064114 | |
| UN2922 | |
| Air sensitive | |
| RTCUCQWIICFPOD-SECBINFHSA-N | |
| (1R)-1-naphthalen-1-ylethanamine | |
| 2724264 | |
| 99% |
Safety and Handling
GHS H Statement
H301-H314-H318-H312
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 223-425-5
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only