missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1-(2-Bromophenyl)ethanol, 98%

CAS: 76116-20-6 | C8H9BrO | 201.063 g/mol

$102.38 - $342.47

Chemical Identifiers

CAS 76116-20-6
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00216591
InChI Key DZLZSFZSPIUINR-ZCFIWIBFSA-N
Synonym r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethan-1-ol, pubchem5686, r-+-1-2-bromophenyl ethanol, r-+-2-bromo-, a-methylbenzyl alcohol, benzenemethanol,2-bromo-a-methyl-, ar, r-+-2-bromo-alpha-methylbenzyl alcohol, r-+-2-bromo-alpha-methylbenzyl alcoh
PubChem CID 2734868
IUPAC Name (1R)-1-(2-bromophenyl)ethanol
SMILES CC(C1=CC=CC=C1Br)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5600303
View Documents
Thermo Scientific Chemicals
H5600303
1 g
Each for $102.38
Only null left
 
AAH5600306
View Documents
Thermo Scientific Chemicals
H5600306
5 g
Each for $342.47
Only null left
 
Description

Description

It is the most important dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is the most important dyestuff intermediate.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

76116-20-6
201.063
DZLZSFZSPIUINR-ZCFIWIBFSA-N
2734868
CC(C1=CC=CC=C1Br)O
C8H9BrO
MFCD00216591
r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethan-1-ol, pubchem5686, r-+-1-2-bromophenyl ethanol, r-+-2-bromo-, a-methylbenzyl alcohol, benzenemethanol,2-bromo-a-methyl-, ar, r-+-2-bromo-alpha-methylbenzyl alcohol, r-+-2-bromo-alpha-methylbenzyl alcoh
(1R)-1-(2-bromophenyl)ethanol

Specifications

76116-20-6
C8H9BrO
1 g
DZLZSFZSPIUINR-ZCFIWIBFSA-N
CC(C1=CC=CC=C1Br)O
201.063
201.06
(R)-(+)-1-(2-Bromophenyl)ethanol
54°C to 56°C
MFCD00216591
r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethanol, 1r-1-2-bromophenyl ethan-1-ol, pubchem5686, r-+-1-2-bromophenyl ethanol, r-+-2-bromo-, a-methylbenzyl alcohol, benzenemethanol,2-bromo-a-methyl-, ar, r-+-2-bromo-alpha-methylbenzyl alcohol, r-+-2-bromo-alpha-methylbenzyl alcoh
+54° (c=1 in Chloroform)
(1R)-1-(2-bromophenyl)ethanol
2734868
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only