Learn More
(R)-(+)-1-(3-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals
$869.53 - $869.53
Chemical Identifiers
CAS | 88196-70-7 |
---|---|
Molecular Formula | C9H13NO |
Molecular Weight (g/mol) | 151.209 |
MDL Number | MFCD00671655 |
InChI Key | CJWGCBRQAHCVHW-SSDOTTSWSA-N |
Synonym | r-1-3-methoxyphenyl ethanamine, r-1-3-methoxyphenyl ethylamine, 1-r-3-methoxyphenol ethylamine, 1r-1-3-methoxyphenyl ethanamine, 1r-1-3-methoxyphenyl ethan-1-amine, r-+-1-3-methoxyphenyl ethylamine, phenol,3-1r-1-aminoethyl, 1r-1-3-methoxyphenyl ethylamine, r-3-methoxy-alpha-methylbenzylamine, r-1 3-methoxyphenyl ethylamine |
PubChem CID | 7020759 |
IUPAC Name | (1R)-1-(3-methoxyphenyl)ethanamine |
SMILES | CC(C1=CC(=CC=C1)OC)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1632314
|
Thermo Scientific Chemicals
L1632314 |
25 g |
Each for $869.53
|
|
|||||
Description
(R)-(+)-1-(3-Methoxyphenyl)ethylamine is used in dietary supplements and cosmeticeuticals. It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(+)-1-(3-Methoxyphenyl)ethylamine is used in dietary supplements and cosmeticeuticals. It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in water (10g/L at 20°C).
Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. It is sensitive to air. Store under dry inert gas. Incompatible with oxidizing agents.
Chemical Identifiers
88196-70-7 | |
151.209 | |
CJWGCBRQAHCVHW-SSDOTTSWSA-N | |
7020759 | |
CC(C1=CC(=CC=C1)OC)N |
C9H13NO | |
MFCD00671655 | |
r-1-3-methoxyphenyl ethanamine, r-1-3-methoxyphenyl ethylamine, 1-r-3-methoxyphenol ethylamine, 1r-1-3-methoxyphenyl ethanamine, 1r-1-3-methoxyphenyl ethan-1-amine, r-+-1-3-methoxyphenyl ethylamine, phenol,3-1r-1-aminoethyl, 1r-1-3-methoxyphenyl ethylamine, r-3-methoxy-alpha-methylbenzylamine, r-1 3-methoxyphenyl ethylamine | |
(1R)-1-(3-methoxyphenyl)ethanamine |
Specifications
88196-70-7 | |
1.023 | |
>100°C (212°F) | |
1.5335 | |
25 g | |
6683688 | |
r-1-3-methoxyphenyl ethanamine, r-1-3-methoxyphenyl ethylamine, 1-r-3-methoxyphenol ethylamine, 1r-1-3-methoxyphenyl ethanamine, 1r-1-3-methoxyphenyl ethan-1-amine, r-+-1-3-methoxyphenyl ethylamine, phenol,3-1r-1-aminoethyl, 1r-1-3-methoxyphenyl ethylamine, r-3-methoxy-alpha-methylbenzylamine, r-1 3-methoxyphenyl ethylamine | |
CJWGCBRQAHCVHW-SSDOTTSWSA-N | |
CC(C1=CC(=CC=C1)OC)N | |
151.209 | |
151.21 | |
(R)-(+)-1-(3-Methoxyphenyl)ethylamine |
5°C | |
66°C (0.38 mmHg) | |
C9H13NO | |
MFCD00671655 | |
UN2735 | |
Air Sensitive | |
Soluble in water (10g/L at 20°C). | |
+27° (Neat) | |
(1R)-1-(3-methoxyphenyl)ethanamine | |
7020759 | |
≥99% | |
ee 98% |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c
H302-H314-H317-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 618-126-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only