missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine 95.0+%, TCI America™
Supplier: TCI America A13371G
Specifications
(R)-(+)-1-Amino-2-(methoxymethyl)pyrrolidine | |
Yellow | |
MFCD00010622 | |
r-+-1-amino-2-methoxymethyl pyrrolidine, r-2-methoxymethyl pyrrolidin-1-amine, 1-pyrrolidinamine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl pyrrolidin-1-amine, r-1-amino-2-methoxymethyl pyrrolidine, r-1-+ amino-2-methoxymethylpyrrolidine, 2-methoxymethyl-1-pyrrolidinamine #, pubchem8114, r-1-amino-2-methoxymethylpyrrolidine, 2r-2-methoxymethyl pyrrolidinylamine | |
COCC1CCCN1N | |
130.191 | |
130.19 | |
Liquid |
72748-99-3 | |
C6H14N2O | |
1 g | |
BWSIKGOGLDNQBZ-ZCFIWIBFSA-N | |
(2R)-2-(methoxymethyl)pyrrolidin-1-amine | |
2724538 | |
≥95.0% (GC) |
Chemical Identifiers
72748-99-3 | |
130.191 | |
BWSIKGOGLDNQBZ-ZCFIWIBFSA-N | |
2724538 | |
COCC1CCCN1N |
C6H14N2O | |
MFCD00010622 | |
r-+-1-amino-2-methoxymethyl pyrrolidine, r-2-methoxymethyl pyrrolidin-1-amine, 1-pyrrolidinamine, 2-methoxymethyl-, 2r, 2r-2-methoxymethyl pyrrolidin-1-amine, r-1-amino-2-methoxymethyl pyrrolidine, r-1-+ amino-2-methoxymethylpyrrolidine, 2-methoxymethyl-1-pyrrolidinamine #, pubchem8114, r-1-amino-2-methoxymethylpyrrolidine, 2r-2-methoxymethyl pyrrolidinylamine | |
(2R)-2-(methoxymethyl)pyrrolidin-1-amine |
Safety and Handling
TSCA : No