missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1-Methyl-3-phenylpropylamine, 98%, Thermo Scientific™

Chemical Identifiers

CAS 937-52-0
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00145208
InChI Key WECUIGDEWBNQJJ-UHFFFAOYNA-N
Synonym r---1-methyl-3-phenylpropylamine, r-4-phenylbutan-2-amine, 2r-4-phenylbutan-2-amine, unii-o4f6h42uwq, o4f6h42uwq, r-1-methyl-3-phenylpropylamine, r-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, r, r-1-methyl-3-phenyl-propylamine
PubChem CID 2734033
IUPAC Name 4-phenylbutan-2-amine
SMILES CC(N)CCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC414760010
SDS View Documents
Thermo Scientific Chemicals
414760010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC414760050
SDS View Documents
Thermo Scientific Chemicals
414760050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

937-52-0
149.24
WECUIGDEWBNQJJ-UHFFFAOYNA-N
2734033
CC(N)CCC1=CC=CC=C1
C10H15N
MFCD00145208
r---1-methyl-3-phenylpropylamine, r-4-phenylbutan-2-amine, 2r-4-phenylbutan-2-amine, unii-o4f6h42uwq, o4f6h42uwq, r-1-methyl-3-phenylpropylamine, r-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, r, r-1-methyl-3-phenyl-propylamine
4-phenylbutan-2-amine

Specifications

937-52-0
Authentic
Glass bottle
1.5130 to 1.5150
1g
− 11.00 (20.00°C neat)
r---1-methyl-3-phenylpropylamine, r-4-phenylbutan-2-amine, 2r-4-phenylbutan-2-amine, unii-o4f6h42uwq, o4f6h42uwq, r-1-methyl-3-phenylpropylamine, r-4-phenylbutane-2-amine, 4-phenylbutan-2-amine, r, r-1-methyl-3-phenyl-propylamine
CC(N)CCC1=CC=CC=C1
149.24
149.24
(R)-(-)-1-Methyl-3-phenylpropylamine
0.9360g/mL
98%
C10H15N
MFCD00145208
0.936
− 11.00
WECUIGDEWBNQJJ-UHFFFAOYNA-N
4-phenylbutan-2-amine
2734033
98%
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only