missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-1-Phenylethanol, 99%, ee 97+%, Thermo Scientific Chemicals

$213.05 - $794.18

Chemical Identifiers

CAS 1517-69-7
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064263
InChI Key WAPNOHKVXSQRPX-UHFFFAOYNA-N
Synonym r-1-phenylethanol, r-+-1-phenylethanol, 1r-1-phenylethanol, r-alpha-methylbenzyl alcohol, r-+-sec-phenethyl alcohol, 1r-1-phenylethan-1-ol, r-+-1-phenylethyl alcohol, 1-phenylethanol, r, benzenemethanol, alpha-methyl-, r
PubChem CID 637516
ChEBI CHEBI:45616
SMILES CC(O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1929606
View Documents Promotion Details
Thermo Scientific Chemicals
L1929606
5 g
Each for $213.05
 
AAL1929614
View Documents Promotion Details
Thermo Scientific Chemicals
L1929614
25 g
Each for $794.18
 
Description

Description

(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-Phenylethanol is used in the synthesis of optically active products. It is also useful in the determination of enantiomeric purity and for resolutions of acids.

Solubility
Miscible with organic solvents and slightly miscible with water.

Notes
Incompatible with strong acids and strong oxidizing agents.
Specifications

Chemical Identifiers

1517-69-7
122.17
WAPNOHKVXSQRPX-UHFFFAOYNA-N
637516
CC(O)C1=CC=CC=C1
C8H10O
MFCD00064263
r-1-phenylethanol, r-+-1-phenylethanol, 1r-1-phenylethanol, r-alpha-methylbenzyl alcohol, r-+-sec-phenethyl alcohol, 1r-1-phenylethan-1-ol, r-+-1-phenylethyl alcohol, 1-phenylethanol, r, benzenemethanol, alpha-methyl-, r
CHEBI:45616

Specifications

1517-69-7
1.012
85°C (185°F)
C8H10O
MFCD00064263
UN2937
r-1-phenylethanol, r-+-1-phenylethanol, 1r-1-phenylethanol, r-alpha-methylbenzyl alcohol, r-+-sec-phenethyl alcohol, 1r-1-phenylethan-1-ol, r-+-1-phenylethyl alcohol, 1-phenylethanol, r, benzenemethanol, alpha-methyl-, r
WAPNOHKVXSQRPX-UHFFFAOYNA-N
CC(O)C1=CC=CC=C1
122.17
CHEBI:45616
99%
ee ≥97%
9°C to 11°C
88°C to 89°C (10 mmHg)
Mild
1.528
5 g
2039798
Miscible with organic solvents and slightly miscible with water.
+45° (c=5 in Methanol)
1-phenylethan-1-ol
637516
122.17
(R)-(+)-1-Phenylethanol
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H318

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALPHA-METHYLBENZYL ALCOHOL, LIQUID

EINECSNumber : 604-814-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only