Learn More
(R)-2-Aminomethyl-1-Boc-pyrrolidine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5241906
Description
Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
(R)-2-Aminomethyl-1-Boc-pyrrolidine | |
98°C to 110°C (1 mmHg) | |
MFCD03419256 | |
UN2735 | |
Slightly soluble in water. | |
+47° (c=1 in Chloroform) | |
tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate | |
1512534 | |
97% |
259537-92-3 | |
C10H20N2O2 | |
5 g | |
r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidine | |
SOGXYCNKQQJEED-MRVPVSSYSA-N | |
CC(C)(C)OC(=O)N1CCCC1CN | |
200.282 | |
200.28 |
Chemical Identifiers
259537-92-3 | |
200.282 | |
SOGXYCNKQQJEED-MRVPVSSYSA-N | |
1512534 | |
CC(C)(C)OC(=O)N1CCCC1CN |
C10H20N2O2 | |
MFCD03419256 | |
r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidine | |
tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only