Learn More
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid, Thermo Scientific Chemicals
Reagent for hepatic glutathione studies
$91.34 - $91.34
Chemical Identifiers
CAS | 19771-63-2 |
---|---|
Molecular Formula | C4H5NO3S |
Molecular Weight (g/mol) | 147.148 |
MDL Number | MFCD00066092 |
InChI Key | BMLMGCPTLHPWPY-REOHCLBHSA-N |
Synonym | procysteine, r-2-oxothiazolidine-4-carboxylic acid, 4r-2-oxo-1,3-thiazolidine-4-carboxylic acid, l-2-oxothiazolidine-4-carboxylate, oxothiazolidinecarboxylic acid, l-2-oxothiazolidine-4-carboxylic acid, 4-thiazolidinecarboxylic acid, 2-oxo-, 4r, ccris 7672, oxothiazolidine carboxylate, l, r---2-oxothiazolidine-4-carboxylic acid |
PubChem CID | 72390 |
IUPAC Name | (4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid |
SMILES | C1C(NC(=O)S1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6225403
|
Thermo Scientific Chemicals
J6225403 |
1 g |
Each for $91.34
|
|
|||||
Description
Reagent for hepatic glutathione studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
19771-63-2 | |
147.148 | |
BMLMGCPTLHPWPY-REOHCLBHSA-N | |
72390 | |
C1C(NC(=O)S1)C(=O)O |
C4H5NO3S | |
MFCD00066092 | |
procysteine, r-2-oxothiazolidine-4-carboxylic acid, 4r-2-oxo-1,3-thiazolidine-4-carboxylic acid, l-2-oxothiazolidine-4-carboxylate, oxothiazolidinecarboxylic acid, l-2-oxothiazolidine-4-carboxylic acid, 4-thiazolidinecarboxylic acid, 2-oxo-, 4r, ccris 7672, oxothiazolidine carboxylate, l, r---2-oxothiazolidine-4-carboxylic acid | |
(4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid |
Specifications
174°C (decomposition) | |
White | |
MFCD00066092 | |
14,6950 | |
Soluble in water | |
C1C(NC(=O)S1)C(=O)O | |
147.148 | |
147.15 | |
Powder |
19771-63-2 | |
C4H5NO3S | |
4179169 | |
procysteine, r-2-oxothiazolidine-4-carboxylic acid, 4r-2-oxo-1,3-thiazolidine-4-carboxylic acid, l-2-oxothiazolidine-4-carboxylate, oxothiazolidinecarboxylic acid, l-2-oxothiazolidine-4-carboxylic acid, 4-thiazolidinecarboxylic acid, 2-oxo-, 4r, ccris 7672, oxothiazolidine carboxylate, l, r---2-oxothiazolidine-4-carboxylic acid | |
BMLMGCPTLHPWPY-REOHCLBHSA-N | |
(4R)-2-oxo-1,3-thiazolidine-4-carboxylic acid | |
72390 | |
1 g | |
(R)-(-)-2-Oxothiazolidine-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : XJ5426650
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only