missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-2-Phenylpropionic acid, 97%, Thermo Scientific Chemicals

$110.25 - $303.85

Chemical Identifiers

CAS 7782-26-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00063140
InChI Key YPGCWEMNNLXISK-SSDOTTSWSA-N
Synonym r---2-phenylpropionic acid, r-2-phenylpropanoic acid, 2r-2-phenylpropanoic acid, r-hydratropic acid, --hydratropic acid, r---hydratropic acid, r---2-phenylpropionicacid, r-2-phenylpropionic acid, r-2-phenyl-propionic acid, r-2-phenyl-proprionic acid
PubChem CID 446626
ChEBI CHEBI:43035
IUPAC Name (2R)-2-phenylpropanoic acid
SMILES CC(C1=CC=CC=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1763901
View Documents
Thermo Scientific Chemicals
A1763901
0.25 g
Each for $110.25
 
AAA1763903
View Documents
Thermo Scientific Chemicals
A1763903
1 g
Each for $303.85
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7782-26-5
150.177
YPGCWEMNNLXISK-SSDOTTSWSA-N
446626
(2R)-2-phenylpropanoic acid
C9H10O2
MFCD00063140
r---2-phenylpropionic acid, r-2-phenylpropanoic acid, 2r-2-phenylpropanoic acid, r-hydratropic acid, --hydratropic acid, r---hydratropic acid, r---2-phenylpropionicacid, r-2-phenylpropionic acid, r-2-phenyl-propionic acid, r-2-phenyl-proprionic acid
CHEBI:43035
CC(C1=CC=CC=C1)C(=O)O

Specifications

7782-26-5
1.1
148°C (298°F)
1.523
0.25 g
r---2-phenylpropionic acid, r-2-phenylpropanoic acid, 2r-2-phenylpropanoic acid, r-hydratropic acid, --hydratropic acid, r---hydratropic acid, r---2-phenylpropionicacid, r-2-phenylpropionic acid, r-2-phenyl-propionic acid, r-2-phenyl-proprionic acid
−73° (c=1.6 in Chloroform)
(2R)-2-phenylpropanoic acid
446626
150.18
(R)-(-)-2-Phenylpropionic acid
32°C to 34°C
264°C to 266°C
C9H10O2
MFCD00063140
2207688
YPGCWEMNNLXISK-SSDOTTSWSA-N
CC(C1=CC=CC=C1)C(=O)O
150.177
CHEBI:43035
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only